Learn More
Aviva Systems Biology Corp TSPAN4 Antibody - middle region (ARP44756_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44756_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TSPAN4. Alternate names: NAG-2, NAG2, TETRASPAN, TM4SF7, TSPAN-4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-63056