Learn More
Aviva Systems Biology Corp TSPAN5 Antibody - middle region (ARP46640_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPAN5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TSPAN5. Alternate names: NET-4, TM4SF9, TSPAN-5, NET4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53482