Learn More
New England Biolabs, Inc. TspMI – 200 units
SDP
Catalog No. 501526447
Encompass
A restriction endonuclease that recognizes the sequence C^CCGG_G.
Catalog No. 50-152-6447 Supplier New England Biolabs, Inc. Supplier No. R0709S
A restriction endonuclease that recognizes the sequence C^CCGG_G.