Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TSPP tetrasodium | 39050-26-5 | MFCD00064854 | 95.0% | 1022.92 | C44H26N4Na4O12S4 | 250 MG
SDP

Catalog No. 5000304082
Encompass_Preferred

TSPP tetrasodium is a water-soluble porphyrin-based photosensitizer for photodynamic research and fluorescence studies. It is supplied as a light-sensitive solid suitable for laboratory use and exhibits distinct excitation and emission properties useful in imaging and antimicrobial investigations.

  • serves as a porphyrin-based photosensitizer for photodynamic and antimicrobial research.
  • excitation and emission: Ex 413 nm, Em 640 nm.
  • water-soluble tetrasodium salt form, facilitating aqueous applications.
  • high purity (≈95.0%) suitable for research use.
  • appearance: brown to black solid; protect from light during storage.
  • storage recommendations: 4°C under inert atmosphere; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-04082 Supplier Medchemexpress LLC Supplier No. 250MGHYW035051
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TSPP tetrasodium is a water-soluble porphyrin-based photosensitizer for photodynamic research and fluorescence studies. It is supplied as a light-sensitive solid suitable for laboratory use and exhibits distinct excitation and emission properties useful in imaging and antimicrobial investigations.

  • serves as a porphyrin-based photosensitizer for photodynamic and antimicrobial research.
  • excitation and emission: Ex 413 nm, Em 640 nm.
  • water-soluble tetrasodium salt form, facilitating aqueous applications.
  • high purity (≈95.0%) suitable for research use.
  • appearance: brown to black solid; protect from light during storage.
  • storage recommendations: 4°C under inert atmosphere; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.