Learn More
New England Biolabs, Inc. TspRI – 1000 units
SDP
Catalog No. 50812286
Encompass
A restriction endonuclease that recognizes the sequence _NNCASTGNN^.
Catalog No. 50-812-286 Supplier New England Biolabs, Inc. Supplier No. R0582S
A restriction endonuclease that recognizes the sequence _NNCASTGNN^.