Learn More
Aviva Systems Biology Corp TSPYL6 Antibody - N-terminal region (ARP54380_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54380_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSPYL6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TSPYL6. Alternate names: DKFZp434B125ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-61193