Learn More
Enzo Life Sciences TSQ (ultra pure), (25mg), CAS Number: 109628-27-5
SDP
Catalog No. NC1771514
Encompass_Preferred
Zinc dye . TSQ is an efficient membrane-permeable fluorescent probe for zinc (II). It is selective for Zn (II) in the presence of physiological concentrations of Ca (II) and Mg (II) ions. Fluorescence of TSQ is enhanced upon binding Zn (II), but the probe does not demonstrate a shift or change in emission wavelength. TSQ forms a largely insoluble 2:1 (Ligand-Metal) complex with zinc and emits blue light upon excitation at 365 nanometers. Alternative Name: N-(6-Methoxy-8-quinolyl)-p-toluenesulfonamide. Formula: C17H16N2O3S. MW: 328.4. CAS Number: 109628-27-5. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Use/Stability: Stable for at least one year after receipt when stored as recommended. Long Term Storage: -20°C
Catalog No. NC1771514 Supplier Enzo Life Sciences Supplier No. ENZ52153
Zinc dye . TSQ is an efficient membrane-permeable fluorescent probe for zinc (II). It is selective for Zn (II) in the presence of physiological concentrations of Ca (II) and Mg (II) ions. Fluorescence of TSQ is enhanced upon binding Zn (II), but the probe does not demonstrate a shift or change in emission wavelength. TSQ forms a largely insoluble 2:1 (Ligand-Metal) complex with zinc and emits blue light upon excitation at 365 nanometers. Alternative Name: N-(6-Methoxy-8-quinolyl)-p-toluenesulfonamide. Formula: C17H16N2O3S. MW: 328.4. CAS Number: 109628-27-5. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Use/Stability: Stable for at least one year after receipt when stored as recommended. Long Term Storage: -20°C