Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl TSS Clear Wire Shelf Label Holder

Catalog No. 11222124
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
60 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-222-124 24 in.
11-222-125 36 in.
11-222-126 48 in.
11-222-127 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-222-124 Supplier Wanzl Supplier No. T.LH24CLEAR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wire Shelf Label Holder- Clear for TSS Shelving System

  • Add-on option for additional customization
  • Clear
  • Three ways to mount

Specifications

Includes Wire Shelf Label Holder
Color Clear
Width (English) 24 in.
For Use With (Application) Shelving
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.