Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wanzl TSS Clear Wire Shelf Label Holder

Catalog No. 11222127
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
60 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
11-222-127 60 in.
11-222-124 24 in.
11-222-125 36 in.
11-222-126 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 11-222-127 Supplier Wanzl Supplier No. T.LH60CLEAR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Wire Shelf Label Holder- Clear for TSS Shelving System

  • Add-on option for additional customization
  • Clear
  • Three ways to mount

Specifications

Color Clear
For Use With (Application) Shelving
Width (English) 60 in.
Includes Wire Shelf Label Holder
Material Plastic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.