Learn More
Hach Company Glass Fiber Filter, Diameter 47 mm, Preweighed for Standard Methods TSS procedure, 400/pk
Supplier:  Hach Company 2546200
Encompass
Individually preweighed filter papers, preweighed in compliance with Standard Methods Total Suspended Solids procedure. Each in a planchet with tare weight listed. Each tare weight meets Standard Methods specifications.
Catalog No. NC2018156