Learn More
Aqua Solutions TSS Standard 20 mg/L (Celite - Shake bottle well before use) (1L)
SDP
Catalog No. NC0838368
Encompass
TSS Standard 20 mg/L (Celite - Shake bottle well before use) (1L)
Catalog No. NC0838368 Supplier Aqua Solutions Supplier No. EQS513
TSS Standard 20 mg/L (Celite - Shake bottle well before use) (1L)