Learn More
IW Tremont Co Inc LabExact Filter Binderless glass microfiber TSS 1.5um 47cm dia. Use for EPA Methods 2540C and 2540D prewashed & dried, not tared, TSS Procedure, testing dissolved and suspended solids in water gravimetric analysis of air pollutants 100/pk
SDP
Supplier:  IW Tremont Co Inc LETSSWD4700
Encompass_Preferred
LabExact Pre-washed and dried IW Tremont Grade TSS filters for Total Suspended Solids (TSS) are processed using reagent grade water (RGW) under strict controls to wash away extraneous fiber per the requirements of the EPA Standard 2540 C, D & E. Prepared filters are an effective time saver that removes a step from your work flow while maintaining your high levels of quality assurance. Filters are packaged in plastic container, in stacks without weighing pans or bar coding. Available in both 47 and 90mm diameters. Package of 100 washed and ready to use filters
Catalog No. 50-223-7309