Learn More
Carnabio Usa Inc TSSK2/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 021655
Encompass
TSSK2, Full-length, Wild type, Amino Acid 1-358, Q96PF2, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0649