Learn More
Aviva Systems Biology Corp TSSK2 Antibody - middle region (ARP53819_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53819_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TSSK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human TSSK2. Alternate names: DGS-G, FLJ38613, SPOGA2, STK22B, TSK2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-893