Learn More
Hach Company Free & Total Chlorine Test Strips, 0-10 mg/L, Individually Wrapped, 250/pk
Catalog No. 501996681 Shop All Hach Company Products
Encompass
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Chlorine exists in water as "free" chlorine (hypochlorite) or "combined" chlorine (chloramines). The total chlorine test detects both forms. With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes, and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing. Strips are individually wrapped for maximum portability and stability. Contains 250 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6681 Supplier Hach Company Supplier No. 2793944
The most widely used disinfectant for drinking water, chlorine is also important for sanitizing swimming pools, cooling towers, other industrial equipment, and municipal wastewater. Its measurement and control are vital for both safety and economic reasons. Chlorine exists in water as "free" chlorine (hypochlorite) or "combined" chlorine (chloramines). The total chlorine test detects both forms. With Hach Water Quality Test Strips, technicians in the field can test many samples in only a few minutes, and make immediate evaluations on-site. No measuring, set-up, clean-up, or chemical handling are necessary. Hach test strips are also used in laboratories all over the world for pre-test screening-to detect the presence of materials that might interfere with lab testing. Strips are individually wrapped for maximum portability and stability. Contains 250 tests.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.