Learn More
Grainger Test Strips, Peroxide, 1-100 ppm, 50 pack
Catalog No. NC9983450
Encompass_Preferred
Peroxide Strip, 0 to 100 ppm, Test On-Site for a Variety of Metals, pH, etc.
Catalog No. NC9983450 Supplier Grainger Supplier No. 3VEW7
Peroxide Strip, 0 to 100 ppm, Test On-Site for a Variety of Metals, pH, etc.