Learn More
Research Products International Corp Test Tube pH Pen, Flat Tip Probe, ATC
SDP
Catalog No. 502229870
Encompass
Highly stable and accurate readings with a large LCD display. Designed for simple one-hand operation. Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize any buffer. Features hold function, and auto power off with low battery indicator. Variety of probes available for all applications:  Flat Tip Probe for surface measurement. The case is IP65 waterproof and will float if accidentally dropped into the water. Supplied with 2 button-cell batteries and a built-in electrode that is kept moist with a protective cap.
Catalog No. 50-222-9870 Supplier Research Products International Corp Supplier No. 850066
Highly stable and accurate readings with a large LCD display. Designed for simple one-hand operation. Automatically calibrates to pH 4, 7 and 10 buffers and can be adjusted to recognize any buffer. Features hold function, and auto power off with low battery indicator. Variety of probes available for all applications:  Flat Tip Probe for surface measurement. The case is IP65 waterproof and will float if accidentally dropped into the water. Supplied with 2 button-cell batteries and a built-in electrode that is kept moist with a protective cap.