Learn More
US Plastic Corporation 15mL Kartell Polypropylene Test Tube with Blue Screw Closure, 100/cs
SDP
Catalog No. NC1751060
Encompass
15ml test tubes with polyethylene quick closure screw caps. The design of the cap avoids any contamination due to drops of liquid on the inner surface.
Catalog No. NC1751060 Supplier US Plastic Corporation Supplier No. 77426
15ml test tubes with polyethylene quick closure screw caps. The design of the cap avoids any contamination due to drops of liquid on the inner surface.