Learn More
Electron Microscopy Sciences T-Sue™ Microarray Mold Kit, 1 mm
SDP
Catalog No. 501930038
Encompass_Preferred
T-Sue array molds are specifically designed to be simple, easy to use and inexpensive. The tissues can be analyzed in consistent conditions enhancing the efficiency of the research. Allow you to perform TMAs more quickly with excellent results. By using array molds, you can process up to 170 specimens onto one single slide in very little time.
Made of silicone, these molds can withstand temperatures between -100 deg C to +250 deg C and be reused hundreds of times without losing their flexibility.
Each kit includes one T-Sue array mold and four punch needles with stylet.
Size: 1 mm
Capacity: 170 cores
Catalog No. 50-193-0038 Supplier Electron Microscopy Sciences Supplier No. 6913207
T-Sue array molds are specifically designed to be simple, easy to use and inexpensive. The tissues can be analyzed in consistent conditions enhancing the efficiency of the research. Allow you to perform TMAs more quickly with excellent results. By using array molds, you can process up to 170 specimens onto one single slide in very little time.
Made of silicone, these molds can withstand temperatures between -100 deg C to +250 deg C and be reused hundreds of times without losing their flexibility.
Each kit includes one T-Sue array mold and four punch needles with stylet.
Size: 1 mm
Capacity: 170 cores