Learn More
Andwin Scientific TSUTEK REPL SCISSOR BLADE 5/CS
SDP
Catalog No. NC9928143
Encompass_Preferred
The Tissue-Tek Accu-Edge replaceable blade is an ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold in a case of 5.
Catalog No. NC9928143 Supplier Andwin Scientific Supplier No. 4795
The Tissue-Tek Accu-Edge replaceable blade is an ideal tool for performing anatomical sectioning of tissues. The blades can be easily inserted and removed, which removes the time, expense, and incovenience of sharpening. This product is autoclavable and is sold in a case of 5.