Catalog No. TSV72RPGAPM
Click to view available options
Cabinet Material:
Painted Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Cabinet Material
TSV72RPGAPM
Painted Steel
TSV72RSGAPM
Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Attention: This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No.
TSV72RPGAPM
Supplier
Thermo Scientific™
Supplier No.
TSV72RPGAPM
Attention: This item requires special delivery information. You will be asked for this during the checkout process.
Thermo Scientific TSV Series 72 cu.ft. lab refrigerators offer a convenient option for storage of non-critical samples and reagents.
Three self-closing glass doors Remote alarm contacts High and low temperature alarms Digital controller with adjustable temperature setpoint Door ajar alarm Optional stainless steel interior
Specifications
Capacity (English)
72 cu. ft.
Capacity (Metric)
2038 L
Temperature Range
4°C to 6°C
Temperature (Setpoint)
5°C
Refrigerant
R290
Door Style
Glass Self-closing with Lock
No. of Doors
3
Shelves
12 Adjustable
Capacity (English) Shelf
100 lb.
Capacity (Metric) Shelf
45 kg
Cabinet Material
Painted Steel
Exterior Finish
Painted Steel
Certifications/Compliance
cULus
Style
Freestanding
Display
Digital
Casters
Yes, customer installed
Ports
One rear access port
Warranty
2 Years Parts and Labor, 3 Additional Years Compressor
Defrost
Automatic
Monitoring Options
SMART VUE Compatible, Remote Alarm Contacts
Access Security
Key Lock
Compressor
Bottom-mounted
Dimensions (DxWxH) Exterior
75.7 x 32 x 81 in. (1924 x 813 x 2057 mm)
Dimensions (D x W x H) Interior
72 x 28.5 x 61.75 in. (1829 x 724 x 1569 mm)
Plug Type
NEMA 5-15
Electrical Requirements
120 V, 60 Hz
Shipping Weight (English)
730 lb.
Shipping Weight (Metric)
318 kg
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?