Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSV Value Combo Refrigerator/Freezer and General Purpose Bath Bundle

Catalog No. TSV18CPSAWB Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
510 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
TSV18CPSAWB 510 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSV18CPSAWB Supplier Thermo Scientific™ Supplier No. TSV18CPSAWB

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Chill, freeze and retrieve samples safely, simply and economically with the TSV Value Refrigerator/Freezer. Includes General Purpose Bath, 10 L.

  • Factory tested prior to shipment
  • NRTL listed
  • Environmentally-friendly, SNAP-compliant hydrocarbon refrigerants
  • Adjustable temperature control
  • Easy to clean, durable finish
  • Manual and auto-defrost combination units
TRUSTED_SUSTAINABILITY

Specifications

Type Combination Refrigerator/Freezer
Capacity (English) 18 ft.3
Capacity (Metric) 510 L
Temperature Range 1°C to 12°C
Refrigerant R600a
Door Style Hinged
No. of Doors 2 (2 compartments)
Hinge Side Right
Shelves 3
Door Shelves 3 Shelves
Exterior Finish Enameled Steel
Insulation Cyclopentane
Interior ABS
Certifications/Compliance cULus
Casters 2 Leveling Casters
Color White
Control Type Electronic
For Use With (Application) General Storage of Non-critical Samples and Reagents
Typical Lifetime 10 Years
Defrost Automatic
Lighting Interior
Min. Clearance 4 in. (Sides), 4 in. (Back), 11 in. (Top)
Compressor Bottom-mounted
Dimensions (D x W x H) Interior Refrigerator: 64.7 x 63.5 x 101.6 cm (25.5 x 25 x 40 in.), Freezer: 52 x 57.1 x 40 cm (20.5 x 22.5 x 15.75 in.)
Dimensions (L x W x H) Exterior 83.3 x 75 x 169.2 cm (32.79 x 29.53 x 66.61 in.)
Amperage 1.1 A
Plug Type NEMA 5-15 P
Voltage 115 V
Frequency 60 Hz.
Green Features 115 V
Weight (English) 224 lb.
Weight (Metric) 101.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.