Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Assure Series High Performance Manual Defrost Laboratory Freezer

Catalog No. TSX23020FV Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
418 L
698 L
818 L
Voltage:
115 V
220 V
Plug Type:
CEE 7/7
NEMA 5-15
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Voltage Plug Type
TSX23020FV 818 L 220 V CEE 7/7
TSX21520FV 418 L 220 V CEE 7/7
TSX21520FA 418 L 115 V NEMA 5-15
TSX22520FV 698 L 220 V CEE 7/7
TSX22520FA 698 L 115 V NEMA 5-15
TSX23020FA 818 L 115 V NEMA 5-15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. TSX23020FV Supplier Thermo Scientific™ Supplier No. TSX23020FV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ TSX Series high-performance -20°C manual defrost freezers offer a sustainable choice for clinical and laboratory storage

Thermo Scientific™ TSX HP Assure Manual Defrost Freezers are designed for laboratories requiring highly stable frozen storage for temperature-sensitive biological and laboratory materials. Manual defrost operation minimizes temperature variation associated with automatic defrost cycles, while touchscreen controls, onboard data logging with USB download, and the trusted TSX platform provide dependable performance for demanding laboratory applications.

Reliable performance for temperature-sensitive frozen storage

  • Designed for applications requiring stable frozen storage of biological samples, reagents, pharmaceuticals, vaccines, enzymes, and other temperature-sensitive laboratory materials
  • Ideal for research, clinical, diagnostic, pharmaceutical, biotechnology, industrial, and academic laboratory environments
  • Purpose-built for laboratories where stable frozen storage is critical to sample integrity
  • Designed to deliver dependable performance for demanding laboratory workflows

Designed to protect valuable samples

  • Manual defrost operation supports workflows that are sensitive to the intermittent temperature variation associated with automatic defrost cycles
  • Cold-wall convection cooling supports consistent temperature performance and temperature uniformity
  • Designed to help maintain stable frozen storage conditions for sensitive biological and laboratory materials
  • Built on the trusted Thermo Scientific™ TSX platform to deliver dependable long-term performance and reliability

Designed for confidence at the point of use

  • Modern touchscreen interface simplifies daily operation, configuration, and status review
  • Onboard data logging with USB download provides convenient access to temperature records for monitoring and documentation
  • Intuitive controls support faster training, easier setup, and greater confidence during routine operation
  • Designed to simplify monitoring and documentation without adding complexity to laboratory workflows

Compliance-ready cold storage

  • Designed to support qualification, validation, and audit-ready laboratory environments
  • cULus or CE listed, as appropriate by voltage and region
  • REACH compliant
  • RoHS compliant
  • Proposition 65 compliant
  • WEEE compliant

Sustainable by design

  • Variable-speed compressor technology continuously adapts cooling performance based on laboratory demand while maintaining stable temperature performance
  • Designed to support compliance with applicable European Union F-Gas regulations and the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP)
  • Designed to help laboratories reduce energy consumption without compromising sample protection or performance

Built for today's laboratory

  • Combines trusted TSX performance with improved usability, strong value, and product availability aligned to the pace of modern laboratory work
  • Available as part of the TSX HP Assure application-specific portfolio for laboratories with specialized frozen storage requirements
  • Designed to deliver the performance, compliance confidence, availability, and value laboratories expect

Services and support

  • Compatible with Thermo Fisher Scientific service offerings including preventive maintenance, calibration, qualification, temperature mapping, and regulatory assurance services
  • Supports remote monitoring solutions for centralized asset management and 24/7 monitoring of compatible equipment
  • Service availability may vary by region; contact your Thermo Fisher Scientific representative for details
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 29 cu.ft.
Capacity (Metric) 818 L
Temperature Range -25°C to -15°C
Door Style Solid
No. of Doors 1
Shelves 5
Certifications/Compliance CE, cULus, NSF 456, Prop 65, REACH
Style Upright
Control Type Touch Screen
Defrost Manual
Lighting LED
Dimensions (DxWxH) Exterior 950 x 921 x 1979 mm
Dimensions (D x W x H) Interior 756 x 750 x 1460 mm
Access Security Key Lock
Data Outputs USB
Plug Type CEE 7/7
Voltage 220 V
Frequency 50/60Hz
Compressor V-drive
Weight (English) 518 lb.
Weight (Metric) 235 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.