Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Flex Series High Performance Auto Defrost Laboratory Freezer

Catalog No. TSX12530FV Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
350 L
700 L
Voltage:
115 V
220 V
Plug Type:
CEE 7/7
NEMA 5-15
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Voltage Plug Type
TSX12530FV 700 L 220 V CEE 7/7
TSX11230FV 350 L 220 V CEE 7/7
TSX11230FA 350 L 115 V CEE 7/7
TSX12530FA 700 L 115 V NEMA 5-15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. TSX12530FV Supplier Thermo Scientific™ Supplier No. TSX12530FV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ TSX Series high-performance -30°C auto defrost freezers offer a sustainable choice for clinical and laboratory storage.

Thermo Scientific™ TSX HP Flex Auto Defrost Laboratory Freezers are designed for laboratories requiring dependable frozen storage of vaccines, pharmaceuticals, reagents, biological samples, and other commonly used laboratory materials. Engineered for demanding laboratory workflows, these freezers combine consistent temperature performance, automatic defrost operation, intuitive touchscreen controls, and onboard data logging with USB download to simplify everyday operation while protecting valuable samples.

Reliable performance for everyday laboratory workflows

  • Designed for the storage of vaccines, pharmaceuticals, reagents, biological samples, and other commonly used laboratory materials requiring dependable frozen storage
  • Ideal for research, clinical, diagnostic, pharmaceutical, biotechnology, industrial, and academic laboratory environments
  • Engineered to deliver consistent temperature performance and rapid recovery following routine door openings
  • Purpose-built to help protect valuable samples while simplifying day-to-day laboratory operation

Designed to protect valuable samples

  • Automatic defrost operation minimizes routine maintenance while supporting reliable frozen storage performance
  • Consistent temperature control helps protect temperature-sensitive materials during demanding laboratory workflows
  • Built on the trusted Thermo Scientific™ TSX platform to deliver dependable long-term performance and reliability
  • Designed to balance sample protection, usability, and operational efficiency

Designed for confidence at the point of use

  • Modern touchscreen interface simplifies daily operation, configuration, and status review
  • Onboard data logging with USB download provides convenient access to temperature records for monitoring and documentation
  • Intuitive controls support faster training, easier setup, and greater confidence during routine operation
  • Designed to simplify monitoring and documentation without adding complexity to laboratory workflows

Compliance-ready cold storage

  • Designed to support qualification, validation, and audit-ready laboratory environments
  • cULus or CE listed, as appropriate by voltage and region
  • REACH compliant
  • RoHS compliant
  • Proposition 65 compliant
  • WEEE compliant

Sustainable by design

  • Variable-speed compressor technology continuously adapts cooling performance based on laboratory demand while maintaining stable temperature performance
  • Designed to support compliance with applicable European Union F-Gas regulations and the U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP)
  • Designed to help laboratories reduce energy consumption without compromising sample protection or performance

Built for today's laboratory

  • Combines trusted TSX performance with improved usability, strong value, and product availability aligned to the pace of modern laboratory work
  • Supports laboratories requiring dependable cold storage for routine and regulated environments
  • Designed to deliver the performance, compliance confidence, availability, and value laboratories expect

Services and support

  • Compatible with Thermo Fisher Scientific service offerings including preventive maintenance, calibration, qualification, temperature mapping, and regulatory assurance services
  • Supports remote monitoring solutions for centralized asset management and 24/7 monitoring of compatible equipment
  • Service availability may vary by region; contact your Thermo Fisher Scientific representative for details
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 25 cu.ft.
Capacity (Metric) 700 L
Temperature Range -35°C to -15°C
No. of Doors 1
Shelves 5
Certifications/Compliance CE, cULus, NSF 456, Prop 65, REACH
Style Upright
Control Type Touch Screen
Defrost Automatic
Lighting LED
Dimensions (DxWxH) Exterior 955 x 875 x 1965 mm
Dimensions (D x W x H) Interior 755 x 725 x 1350 mm
Access Security Key Lock
Data Outputs USB
Plug Type CEE 7/7
Voltage 220 V
Frequency 50/60Hz
Compressor V-drive
Weight (English) 463 lb.
Weight (Metric) 210 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.