Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Flex Series High Performance Laboratory Refrigerators

Catalog No. TSX13605SV Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
341 L
1031 L
Door Style:
Glass
Solid
Voltage:
115 V
220 V
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Door Style Voltage
TSX13605SV 1031 L Solid 220 V
TSX11205GV 341 L Glass 220 V
TSX11205SV 341 L Solid 220 V
TSX11205GA 341 L Glass 115 V
TSX11205SA 341 L Solid 115 V
TSX13605GV 1031 L Glass 220 V
TSX13605GA 1031 L Glass 115 V
TSX13605SA 1031 L Solid 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. TSX13605SV Supplier Thermo Scientific™ Supplier No. TSX13605SV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thermo Scientific™ TSX Series high-performance, Innovative refrigeration solutions for healthcare and research needs.

Thermo Scientific™ TSX HP Flex Laboratory Refrigerators are designed for laboratories requiring reliable storage of reagents, media, samples, kits, and other temperature-sensitive materials. Touchscreen controls, onboard data access, and adjustable interior configurations deliver an intuitive user experience while maintaining high-performance temperature control for everyday laboratory workflows

Reliable performance for everyday laboratory workflows

  • Designed for the storage of reagents, media, samples, kits, and other temperature-sensitive laboratory materials
  • Ideal for routine research, clinical, diagnostic, pharmaceutical, biotechnology, industrial, and academic laboratory environments
  • Designed to support convenient access and dependable storage of temperature-sensitive laboratory materials
  • Engineered to deliver dependable temperature performance for everyday laboratory applications

Designed to protect valuable samples

  • Designed to help maintain thermal stability and support efficient cold storage performance
  • Consistent temperature performance helps protect temperature-sensitive materials during routine laboratory use
  • Adjustable interior configurations help accommodate changing inventory and laboratory storage requirements
  • Designed to support dependable storage and efficient everyday laboratory workflows

Designed for confidence at the point of use

  • Modern touchscreen interface simplifies daily operation, configuration, and status review
  • Onboard data logging with USB download provides convenient access to temperature records for monitoring and documentation
  • Intuitive controls support faster training, easier setup, and greater confidence during daily operation
  • Designed to simplify routine monitoring without adding complexity to laboratory workflows

Compliance-ready cold storage

  • Designed to support qualification, validation, and audit-ready laboratory environments
  • cULus or CE listed, as appropriate by voltage and region
  • REACH, RoHS, Proposition 65, and WEEE compliant
  • North American 115V / 60Hz models are ENERGY STAR™ 2.0 certified, where applicable

Sustainable by design

  • Variable-speed compressor technology adapts cooling performance based on laboratory demand while maintaining stable temperatures
  • Designed to support compliance with EU F-Gas regulations and the U.S. EPA Significant New Alternatives Policy (SNAP)
  • Designed to help laboratories reduce energy consumption without compromising sample protection

Built for today's laboratory

  • Combines trusted TSX performance with improved usability, strong value, and product availability
  • Supports laboratories requiring dependable cold storage for regulated and routine environments
  • Designed to deliver the performance, compliance confidence, availability, and value laboratories expect
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 36 cu.ft.
Capacity (Metric) 1031 L
Temperature Range 2°C to 8°C
Door Style Solid
No. of Doors 1
Exterior Finish Powder-coated Steel
Certifications/Compliance CE, cULus, Prop 65, REACH, REACH
Style Upright
Control Type Touch Screen
Warranty 5 years parts and labor, plus an additional 5 years for compressor parts
Defrost Automatic
Lighting LED
Access Security Key Lock
Data Outputs USB
Compressor V-drive
Dimensions (D x W x H) Interior 590 x 1115 x 1670 mm
Dimensions (L x W x H) Exterior 827 x 1205 x 1987 mm
Plug Type CEE 7/7
Voltage 220 V
Frequency 50/60Hz
Weight (English) 573 lb.
Weight (Metric) 260 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.