Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Series High-Performance -30°C Auto Defrost Freezer PROMO

Catalog No. TSX1230FAPM Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (English):
11.5 cu. ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
TSX1230FAPM 11.5 cu. ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSX1230FAPM Supplier Thermo Scientific™ Supplier No. TSX1230FARP

missing translation for 'validMainframeMsg'

Attention:This item requires special delivery information. You will be asked for this during the checkout process.

The quiet, sustainable choice for -30°C, auto-defrost storage

The drive for sample integrity and security

  • Designed to safely store media, reagents, pharmaceuticals, vaccines, chemotherapy and other laboratory and medical-grade storage requiring a tight temperature performance.
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection.
  • Non-invasive fully adaptive defrost with time and temperature guided cycle.
  • Easy to use microprocessor control with interior light, setpoint security and alarms.
  • Standard door locks provide comfort and assurance.

Enabling compliance

  • Tested and Certified to NSF/ANSI 456 Vaccine Storage standard and supports the US CDC, WHO, and other global vaccine storage guidelines and recommendations.
  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments.
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863.

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip.
  • The controller is angled to help prevent neck strain.
  • Easy to clean and adjust epoxy coated shelves.
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction.
  • Self-closing door with 90° stop to assist with inventory loads.

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life.
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility.

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always and anywhere know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems.

Make it your own

  • Alternate Door swing configuration and door types.
  • Extra and Alternate wire baskets, shelves, and drawers.
  • Free-standing or factory-installed chart recorders.
  • Stainless Steel interior finish.
  • E-Lock Adapter kits.
  • 4-20 milliamp transmitters and other remote alarm modules.
  • Specialty, regional, and country specific plugs.

Services and support

  • Industry Leading 120-month compressor warranty (varies by country, please contact us for details).
  • Optional Certificate of Calibration to an NIST (National Institute of Standards and Technology, formerly National Bureau of Standards) traceable instrument includes a model number and serial number certificate.
  • Optional Validation Testing Programs include in house and field-based temperature mapping and Installation Qualification (IQ) and Operation Qualification (OQ) programs.
  • Optional Factory Acceptance Test results including QC Test Log Sheets and QC Temperature Graphs.
TRUSTED_SUSTAINABILITY

Specifications

Type High-Performance Freezer
Capacity (English) 11.5 cu. ft.
Capacity (Metric) 326 L
Temperature (Setpoint) -35° to -15°C (-30°C)
Door Style Solid
No. of Doors 1
Shelves 4
Certifications/Compliance UL, cUL, NSF 456 (When Purchased with 6916-2 Calibration Certificate)
Display Microprocessor
Product Line TSX Series
Defrost Automatic
Dimensions (DxWxH) Exterior 31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior 31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior 31.1 in.
Depth (English) Interior 21.8 in.
Depth (Metric) Exterior 79.0 cm
Depth (Metric) Interior 55.4 cm
Height (English) Exterior 73.0 in.
Height (English) Interior 52.4 in.
Height (Metric) Exterior 185.4 cm
Height (Metric) Interior 133.1 cm
Width (English) Exterior 24.0 in.
Width (English) Interior 20 in.
Width (Metric) Exterior 61.9 cm
Width (Metric) Interior 50.8 cm
Monitoring Options Wireless
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Amperage 15 A
Plug Type NEMA 5-15
Voltage 115 V
Electrical Requirements 115 V, 60 Hz
Frequency 60 Hz
Green Features Energy efficient
Refrigerant R290
Shipping Weight (English) 335 lb.
Shipping Weight (Metric) 152 kg
Weight (English) 281 lb.
Weight (Metric) 128 kg
Rack Capacity 4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.