Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TSX Series High-Performance Undercounter Lab Refrigerators PROMO

Catalog No. TSX505SASP
Change view
Click to view available options
Door Style:
Glass
Single Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Door Style
TSX505SASP Single Solid
TSX505GASP Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. TSX505SASP Supplier Thermo Scientific™ Supplier No. TSX505SASP/DEL
This item is not returnable. View return policy
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

For a better environment inside and out - Introducing our new undercounter lab refrigerator.

Designed for your environment, new Thermo Scientific™ TSX Series high-performance undercounter lab refrigerators support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other research and medical products. Powered by V-drive, the TSX Series delivers low energy, decreased noise levels and improved performance and user experience.  These new undercounter refrigerators are quiet and feature an improved user interface, automated locking and increased storage capacity and reliability.

Designed to maintain temperature uniformity to protect medical, pharmaceutical-grade products, laboratory media, reagents and more.

  • Unique V-drive technology is designed to save energy without compromising the protection of stored products
  • U.S. Environmental Protection Agency SNAP (Significant New Alternatives Policy) compliant refrigerants
  • Quiet operation at just 35dbA (43 during pull down)
  • Reduced energy consumption
  • Sleek exterior design and touch screen
  • Bright, easy-to-adjust interior LED lighting
  • Touchscreen controller with password protection
  • Datalogging with USB download functionality
  • Battery backup
  • Choice of reversible glass or solid door
  • Air or glycol temperature display
  • Automated locking
  • 1-inch rear access port
  • Remote alarm contacts
  • Audible and visual alarms: power fail, high temp, low temp, low battery, memory full, door ajar
  • ADA compliant
  • Compatible with most major electronic locks
TRUSTED_SUSTAINABILITY

Specifications

Description TSX Series High-Performance Undercounter Refrigerator, Solid Door
Type High-Performance Refrigerator
Capacity (English) 5.5 cu. ft.
Capacity (Metric) 156 L
Temperature Range 3°C to 7°C
Refrigerant R744
Door Style Single Solid
Shelves 3
Certifications/Compliance CE, REACH, RoHS, UL
Display Full color TFT-LCD; 4.3in; 480 x 272
Product Line TSX Series
Defrost Automatic
Smart View Compatibility Smart-Vue Compatible
Dimensions (D x W x H) Interior 20.5 x 19.5 x 23 in. (52 x 49.5 x 58.4 cm)
Dimensions (L x W x H) Exterior 26 x 23.6 x 31.8 in. (66 x 60 x 81 cm)
Depth (English) Exterior 26 in.
Depth (English) Interior 20.5 in.
Depth (Metric) Exterior 66 cm
Depth (Metric) Interior 52 cm
Height (English) Exterior 31.8 in.
Height (English) Interior 23 in.
Height (Metric) Exterior 81 cm
Height (Metric) Interior 58.4 cm
Width (English) Exterior 23.6 in.
Width (English) Interior 19.5 in.
Width (Metric) Exterior 60 cm
Width (Metric) Interior 49.5 cm
Amperage 10 A Max / 3 A Steady State
Plug Type NEMA 5-15 P
Voltage 100/240 V
Electrical Requirements 100 - 240 V 50-60 Hz
Frequency 50/60 Hz
Shipping Weight (English) 155 lb.
Shipping Weight (Metric) 71 kg
Weight (English) 119 lb.
Weight (Metric) 53.98 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.