Learn More
Thermo Scientific™ TSX Universal Series General Purpose ULT Freezers - Trade-Up Program (2-Inner Doors)
Shop All Thermo Scientific Products

Description
Reliability at its core:
- Manufactured excellence: built to the highest standards at our ISO 13485:2016 facilities
- Proven durability: 10k+ hours of intensive lab and field runtime testing to help ensure long-term reliability
- Proven quality: each unit undergoes rigorous end-of-line testing for quality control
- Complete transparency: shipped with factory documentation and a certification of calibration1
- Peace of mind: backed by our 12-year warranty2 for long-term confidence
Built to perform, designed to protect:
- Enhanced performance with universal V-Drive technology: adapts to user activity to help maintain peak performance and protect what matters most - your samples.
- Stable temperatures: less than 4°C variation helps keep samples safe anywhere in the cabinet
- Fast recovery: industry-leading 15-minute door recovery helps minimize temperature fluctuations and downtime
- Rapid cooling, faster setup: reaches -80°C in under 5 hours - 41% faster than previous models
- Cost effective: uses 33% less energy than previous models , helping reduce costs and environmental impact
- Quick sample access with active pressure relief: innovative pressure relief technology enables re-entry in just 30 seconds so you can retrieve samples as frequently as needed, helping minimize disruptions
- Maximum protection and compact footprint: advanced vacuum insulation and environmentally friendly 0 GWP, water-blown foam insulation safeguard samples while providing a compact footprint
Greener by design
- Less hazardous: uses non-hydrofluorocarbon (HFC) refrigerants and foam insulation which has a 0 GWP and Ozone Depleting Potential (ODP) and is water-blown, helping reduce chemical emissions and outgassing
- Less waste: contains up to 70% recycled materials, with 90% of waste diverted from landfills at our zero-waste facility3, 4
- More energy efficient: ENERGY STAR™ certified4, helping reduce lab energy costs and environmental footprint
- Responsibly packaged: uses non-hydrofluorocarbon (non-HFC) refrigerants with low Global Warming Potential (GWP) and foam insulation, with 0 GWP and Ozone Depleting Potential (ODP), that is water-blown, helping reduce chemical emissions and outgassing
- Extended life: offers a 12-year warranty, longer than any comparable product on the market
Enhanced experience, optimized for ease
- Lab-ready convenience: universal voltage, multiple size options, and whisper-quiet operation for easy installation and compatibility
- Reliable storage and organization: ergonomic handle helps ensure an airtight seal to prevent temperature fluctuations, while adjustable racking maximizes storage efficiency
- Enhanced security and monitoring: 7 in. UI helps streamline operations, optional HID access enhances security, built-in power and outputs enable real-time monitoring and diagnostics, and an optional lightbar allows instant unit status visibility
- Easy frost management: 7-point gasket design with an adjustable perimeter heater helps minimize frost build up for easier maintenance
- Seamless integration: built-in ports for LN2, CO2, and monitoring probes helps ensure compatibility with your lab setup
1Shipped documents:
Certificate of conformance, CE declaration of conformance, certificate of calibration, QC test log sheets, and QC temperature graphs.
2Restrictions and exclusions apply. Warranty is void if the unit is not maintained as set forth in the operation.
3Zero-waste facility in Asheville, North Carolina. Zero waste defined as 90% of non-hazardous waste diverted from landfill, waste to energy and incineration.
4 Information not applicable to TSX Universal ULT freezers with '-ULTS' in the part number (TSX40086FA-ULTS, TSX50086FA-ULTS, TSX60086FA-ULTS.)
Specifications
Specifications
| Type | Ultra-Low Freezer, Upright |
| Capacity (English) | 19.4 cu. ft. |
| Capacity (Metric) | 549 L |
| Temperature Range | -40°C to -86°C |
| Temperature Resolution | 0.1°C |
| Temperature Stability | 0.2°C |
| Door Style | Outer (1), Inner (2) |
| No. of Doors | 1 |
| Door Opening Recovery | 13 (20°C Ambient Running at -75°C) min. |
| Cabinet Material | Scratch-resistant Painted Steel |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.