Learn More
NEOGEN Tetrathionate (TT) Broth Base, Hajna, 500 g
SDP
Catalog No. 502015564
Encompass_Preferred
Tetrathionate (TT) Broth Base, Hajna is used as a selective enrichment for the cultivation of Salmonella species (except S. typhi) from samples in a laboratory setting. Tetrathionate (TT) Broth Base, Hajna is not intended for use in the diagnosis of disease or other conditions in humans. Tetrathionate Broth Base, Hajna, is recommended in the Compendium of Methods for the Microbiological Examination of Foods and the FDA/BAM for the Environmental Sampling and Detection of Salmonella Enteritidis (SE) in Poultry Houses and whole eggs.. Salmonella organisms may be present but injured in food-processing procedures, which include exposure to low temperatures, sub-marginal heat, drying, radiation, preservative, and sanitizers. Numerous reported cases of Salmonella enteritidis (SE) infections are associated with contaminated shell eggs.For more information visit https://www.neogen.com/solutions/Microbiology/tetrathionate-tt-broth-base-hajna
Catalog No. 50-201-5564 Supplier NEOGEN Supplier No. 700003380
Tetrathionate (TT) Broth Base, Hajna is used as a selective enrichment for the cultivation of Salmonella species (except S. typhi) from samples in a laboratory setting. Tetrathionate (TT) Broth Base, Hajna is not intended for use in the diagnosis of disease or other conditions in humans. Tetrathionate Broth Base, Hajna, is recommended in the Compendium of Methods for the Microbiological Examination of Foods and the FDA/BAM for the Environmental Sampling and Detection of Salmonella Enteritidis (SE) in Poultry Houses and whole eggs.. Salmonella organisms may be present but injured in food-processing procedures, which include exposure to low temperatures, sub-marginal heat, drying, radiation, preservative, and sanitizers. Numerous reported cases of Salmonella enteritidis (SE) infections are associated with contaminated shell eggs.For more information visit https://www.neogen.com/solutions/Microbiology/tetrathionate-tt-broth-base-hajna