Learn More
Diversified Biotech Inc TT Cryo-Babies 1.28 x 0.50", 2,000/roll
Catalog No. NC1688260
Encompass
Diversified Biotech patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies and Cryo-Tags also withstand flash/snap freezing in liquid nitrogen.
Catalog No. NC1688260 Supplier Diversified Biotech Inc Supplier No. CRTH1128
Diversified Biotech patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies and Cryo-Tags also withstand flash/snap freezing in liquid nitrogen.