Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellecta Inc All-in-one Tet-Inducible CRISPR Cas9 Lentiviral Vector, Cas9-BFP fusion protein, Doxycycline-inducible, Blasticidin resistance, Packaged Lentiviral Particles, VSV-G pseudotype
SDP

Catalog No. NC2715764
Encompass

Cellecta's product is an all-in-one CRISPR Cas9 lentiviral vector designed for controlled and stable expression of Cas9-BFP for gene editing. This vector allows for the doxycycline-dependent induction of the fusion protein, providing researchers with the flexibility to control gene knockout experiments while minimizing cellular toxicity and off-target effects. It features real-time monitoring of expression and is suitable for various cell types and model organisms.

  • All-in-one lentiviral vector
  • CRISPR Cas9 gene editing system
  • Cas9-BFP fusion for tracking
  • Tetracycline-inducible expression system
  • Includes a blasticidin resistance gene
  • Applicable in both dividing and non-dividing cells
  • Packaged lentiviral particles format
  • Viral pseudotype: VSV-G
  • Storage temperature: -80°C
  • Shelf life: 1 year from receipt

Catalog No. NC2715764 Supplier Cellecta Inc Supplier No. SVRTC9BFPE2BVS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cellecta's product is an all-in-one CRISPR Cas9 lentiviral vector designed for controlled and stable expression of Cas9-BFP for gene editing. This vector allows for the doxycycline-dependent induction of the fusion protein, providing researchers with the flexibility to control gene knockout experiments while minimizing cellular toxicity and off-target effects. It features real-time monitoring of expression and is suitable for various cell types and model organisms.

  • All-in-one lentiviral vector
  • CRISPR Cas9 gene editing system
  • Cas9-BFP fusion for tracking
  • Tetracycline-inducible expression system
  • Includes a blasticidin resistance gene
  • Applicable in both dividing and non-dividing cells
  • Packaged lentiviral particles format
  • Viral pseudotype: VSV-G
  • Storage temperature: -80°C
  • Shelf life: 1 year from receipt

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.