Learn More
Julabo USA Inc Stainless steel test tube rack for 10 Falcon tubes, EA
SDP
Catalog No. 501374460
Encompass_Preferred
Stainless steel test tube rack for 10 Falcon tubes 50 mL, up to +150°C. For use with: CORIO C-B5, CORIO C-BT5, CORIO CD-200F, CORIO CD-201F, CORIO CD-300F, CORIO CD-B5, CORIO CD-BT5, CORIO CP-200F, CORIO CP-201F, CORIO CP-300F, CORIO CP-BC4, CORIO CP-BC6, DYNEO DD-200F, DYNEO DD-201F, DYNEO DD-300F, CORIO CD-BC4, CORIO CD-BC6, DYNEO DD-BC4, DYNEO DD-BC6.
Catalog No. 50-137-4460 Supplier Julabo USA Inc Supplier No. 9970323
Stainless steel test tube rack for 10 Falcon tubes 50 mL, up to +150°C. For use with: CORIO C-B5, CORIO C-BT5, CORIO CD-200F, CORIO CD-201F, CORIO CD-300F, CORIO CD-B5, CORIO CD-BT5, CORIO CP-200F, CORIO CP-201F, CORIO CP-300F, CORIO CP-BC4, CORIO CP-BC6, DYNEO DD-200F, DYNEO DD-201F, DYNEO DD-300F, CORIO CD-BC4, CORIO CD-BC6, DYNEO DD-BC4, DYNEO DD-BC6.