Learn More
Medchemexpress LLC HY-111828 1mg Medchemexpress, TTA-A2 CAS:953778-63-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1118281MG
Encompass_Preferred
Medchemexpress, HY-111828 1mg TTA-A2 CAS:953778-63-7 TTA-A2 is a potent, selective and orally active t-type voltage gated calcium channel antagonist with reduced pregnane X receptor (PXR) activation. TTA-A2 is equally potent against the Cav3.1 (a1G) and Cav3.2 (a1H) channels with IC50 values of 89 nM and 92 nM, respectively, at -80 and -100 mV holding potentials. TTA-A2 can be used for the research of a variety of human neurological diseases, including sleep disorders and epilepsy[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9704