Learn More
Medchemexpress LLC TTA-A2 | 953778-63-7 | 99.33% | 378.39 | 10 MG
SDP
Catalog No. 5000438391
Encompass_Preferred
TTA-A2 is a potent, selective, and orally active T-type voltage-gated calcium channel antagonist with reduced pregnane X receptor (PXR) activation. It is equally potent against the Cav3.1 (a1G) and Cav3.2 (a1H) channels. TTA-A2 can be used for the research of a variety of human neurological diseases, including sleep disorders and epilepsy.
  • Potent and selective T-type voltage-gated calcium channel antagonist
  • Reduced pregnane X receptor activation
  • Equally potent against Cav3.1 and Cav3.2 channels
  • Useful for research into neurological diseases such as sleep disorders and epilepsy
Catalog No. 50-004-38391 Supplier Medchemexpress LLC Supplier No. HY11182810MG
TTA-A2 is a potent, selective, and orally active T-type voltage-gated calcium channel antagonist with reduced pregnane X receptor (PXR) activation. It is equally potent against the Cav3.1 (a1G) and Cav3.2 (a1H) channels. TTA-A2 can be used for the research of a variety of human neurological diseases, including sleep disorders and epilepsy.
  • Potent and selective T-type voltage-gated calcium channel antagonist
  • Reduced pregnane X receptor activation
  • Equally potent against Cav3.1 and Cav3.2 channels
  • Useful for research into neurological diseases such as sleep disorders and epilepsy