Learn More
Medchemexpress LLC TTA-P2 | 1072018-68-8 | 99.6% | 431.37 g/mol | C21H29Cl2FN2O2 | 1 MG
SDP
Catalog No. 5000212674
Encompass_Preferred
TTA-P2 is a research-grade small-molecule inhibitor of T-type (Cav3) calcium channels. It is selective, orally active, and blood-brain barrier permeant, with reported potency (IC50 ≈ 22 nM). Common applications include electrophysiology, studies of neuronal excitability, and in vivo pain and epilepsy models.
  • Selective T-type calcium channel blocker with nanomolar potency.
  • Orally active and blood-brain barrier permeant.
  • Suitable for electrophysiology and in vivo neurological models.
  • Well characterized in pain and epilepsy research literature.
  • Supplied in solid form and in ready-to-use DMSO solution formats.
Catalog No. 50-002-12674 Supplier Medchemexpress LLC Supplier No. HY100351MG
TTA-P2 is a research-grade small-molecule inhibitor of T-type (Cav3) calcium channels. It is selective, orally active, and blood-brain barrier permeant, with reported potency (IC50 ≈ 22 nM). Common applications include electrophysiology, studies of neuronal excitability, and in vivo pain and epilepsy models.
  • Selective T-type calcium channel blocker with nanomolar potency.
  • Orally active and blood-brain barrier permeant.
  • Suitable for electrophysiology and in vivo neurological models.
  • Well characterized in pain and epilepsy research literature.
  • Supplied in solid form and in ready-to-use DMSO solution formats.