Learn More
Medchemexpress LLC HY-10388 10mg Medchemexpress, TTA-Q6 CAS:910484-28-5 Purity:>98%
SDP
Catalog No. 501968172
Encompass_Preferred
Medchemexpress, HY-10388 10mg TTA-Q6 CAS:910484-28-5 TTA-Q6 is a selective T-type Ca2+ channel antagonist, used in the neurological disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8172 Supplier Medchemexpress LLC Supplier No. HY1038810MG
Medchemexpress, HY-10388 10mg TTA-Q6 CAS:910484-28-5 TTA-Q6 is a selective T-type Ca2+ channel antagonist, used in the neurological disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.