Learn More
Medchemexpress LLC Tta-q6 | 910484-28-5 | 100.0% | 405.80 g/mol | C20H15ClF3N3O | 50 MG
SDP
Catalog No. 5000243246
Encompass_Preferred
TTA-Q6 is a selective T-type calcium channel antagonist developed for laboratory research into neurological disease. It shows low-nanomolar potency in depolarized FLIPR assays and sub-micromolar activity in hyperpolarized assays, and is supplied as a high-purity small-molecule research compound for in vitro studies; not for human or veterinary use.
  • Selective T-type calcium channel antagonist.
  • Potent activity in depolarized FLIPR assay (IC50 14 nM).
  • Activity in hyperpolarized FLIPR assay (IC50 590 nM).
  • High purity suitable for research (reported 99.97%).
  • Supplied as a small research pack for laboratory use.
  • Intended for in vitro research only; not for clinical use.
Catalog No. 50-002-43246 Supplier Medchemexpress LLC Supplier No. HY1038850MG
TTA-Q6 is a selective T-type calcium channel antagonist developed for laboratory research into neurological disease. It shows low-nanomolar potency in depolarized FLIPR assays and sub-micromolar activity in hyperpolarized assays, and is supplied as a high-purity small-molecule research compound for in vitro studies; not for human or veterinary use.
  • Selective T-type calcium channel antagonist.
  • Potent activity in depolarized FLIPR assay (IC50 14 nM).
  • Activity in hyperpolarized FLIPR assay (IC50 590 nM).
  • High purity suitable for research (reported 99.97%).
  • Supplied as a small research pack for laboratory use.
  • Intended for in vitro research only; not for clinical use.