Learn More
Med Vet International TTA System Pilot Drill - 2.4 System, 2.5mm Drill Bit Hard, J0372CH
Catalog No. 5000581930
Encompass_Preferred
TTA (Tibial Tuberosity Advancement). TTA is preferred by some surgeons. Further details on the actual surgery can be seen on www.JorVet.com or the instructional DVD provided. This system is available in either titanium or stainless steel; titanium is more expensive but offers better bio-compatibility and fatigue strength.TTA Instrumentation - 2.5mm Drill Bit Hard
Catalog No. 50-005-81930 Supplier Med Vet International Supplier No. JOR372CH
TTA (Tibial Tuberosity Advancement). TTA is preferred by some surgeons. Further details on the actual surgery can be seen on www.JorVet.com or the instructional DVD provided. This system is available in either titanium or stainless steel; titanium is more expensive but offers better bio-compatibility and fatigue strength.TTA Instrumentation - 2.5mm Drill Bit Hard