Learn More
Carnabio Usa Inc TTBK2/10UG/CATALYTIC
Catalog No. 507541017
Encompass
TTBK2, Catalytic domain, Wild type, Amino Acid 1-331, NP_775771.3, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-1017 Supplier Carnabio Usa Inc Supplier No. 0310910
TTBK2, Catalytic domain, Wild type, Amino Acid 1-331, NP_775771.3, Expressed in Insect (sf21); 10microg