Learn More
Camag Scientific Twin Trough Chamber For 20 X 20 Cm Plates, With Stainless Steel Lid
SDP
Catalog No. 501979315
Encompass_Preferred
This Is A Classical Developing Tank For Thin-Layer Chromatography. Compared To A Flat Bottom Chamber Less Solvent Is Needed. A Twin-Trough-Chamber Reduces Disposal Problems And Allows Preconditioning Of The Plate With Any Solvent And For Any Duration. For Preconditioning Or Saturation, The Trough Opposite To The Plate Is Filled With Developing Solvent. The Use Of A Saturation Pad Is Recommended For A Fast, Homogeneous And Reproducible Chamber Saturation Process. This Tank Is Suitable For Plates Up To 20 X 20 Cm In Size.
Catalog No. 50-197-9315 Supplier Camag Scientific Supplier No. 022.5256
This Is A Classical Developing Tank For Thin-Layer Chromatography. Compared To A Flat Bottom Chamber Less Solvent Is Needed. A Twin-Trough-Chamber Reduces Disposal Problems And Allows Preconditioning Of The Plate With Any Solvent And For Any Duration. For Preconditioning Or Saturation, The Trough Opposite To The Plate Is Filled With Developing Solvent. The Use Of A Saturation Pad Is Recommended For A Fast, Homogeneous And Reproducible Chamber Saturation Process. This Tank Is Suitable For Plates Up To 20 X 20 Cm In Size.