Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ TTC/RBS Dipslide (Rose Bengal)

Catalog No. 15398337 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-398-337 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-398-337 Supplier Lovibond™ Supplier No. 56B010310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TTTC/RBS Dipslide (Rose Bengal) is a dual-sided dipslide that detects aerobic bacteria as well as yeast, molds and fungi. Ideal for routine monitoring in petroleum applications. Users can detect growth quickly and can prevent fuel filter blockages and corrosion from acidic conditions in fuel storage tanks.

  • Provides an early indication of bacteria proliferation
  • Effective contact area of 10 cm2 for easy calculation of results
  • Flexible paddle for easy surface testing

A dipslide is an easy-to-use testing solution for microbial content of liquids or surfaces. It contains a sterile culture medium which can be dipped into liquid or pressed on to the surface to collect a sample.

These do-it-yourself tests can be used in the field and deliver semi-quantitative results quickly - with no special training or need to send samples to a laboratory.

When used on a regular basis, users can determine if disinfection protocols are effective and can identity trends over time. Routine monitoring enables early detection of bacteria and other microorganisms proliferating to avoid major issues within the facility or system.

Specifications

Certifications/Compliance ISO 11133
Color Pink
Additive TTC
Detects Bacteria, Yeasts, Molds, and Fungi
Product Type Dip Slide
Target Organism Class Fungi
Formulation Dual Agar with Nutrient TTC Agar and Bengal Red Agar with Chloramphenicol
For Use With (Application) Detect Bacteria in Fuel Systems
For Use With (Equipment) Lovibond Incubator
Incubation Conditions 30°C to 35°C
Incubation Time 24 to 48 hr.
Manual/Automated Manual
No. per Pack 10/Pk.
Packaging Type Pack
Quantity 1 Ea.
Specimen Type Water, Industrial Fluids
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.