Learn More
Aviva Systems Biology Corp TTC12 Antibody - C-terminal region (ARP53725_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53725_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTC12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TTC12. Alternate names: FLJ13859, FLJ20535, TPARMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53727