Learn More
Aviva Systems Biology Corp TTC14 Antibody - N-terminal region (ARP41100_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41100_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTC14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TTC14. Alternate names: DKFZp313M1015, DRDL5813, FLJ00166, KIAA1980, PRO19630ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57723