Learn More
Aviva Systems Biology Corp TTC19 Antibody - C-terminal region (ARP40138_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40138_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TTC19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI505442, 2010204O13Rik, 2810460C24Rik, RP23-330N10.4ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53176