Learn More
Aviva Systems Biology Corp TTC27 Antibody - N-terminal region (ARP57019_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57019_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTC27. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TTC27. Alternate names: FLJ20272ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-254