Learn More
Aviva Systems Biology Corp TTC33 Antibody - N-terminal region (ARP54885_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54885_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TTC33. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TTC33. Alternate names: OSRFApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61353