Learn More
New England Biolabs, Inc. Tte-UvrD Helicase – 0.5 µg
SDP
Catalog No. NC1640071
Encompass
Tte UvrD Helicase is a repair helicase capable of unwinding double-stranded DNA, without a requirement for a specific flap or overhang structure, from the thermophilic organism Thermoanaerobacter tengcongensis. It is active on a wide range of DNA substrates and, along with its thermostability (active to 70C), Tte UvrD Helicase has been demonstrated to be a useful additive for improving specificity of isothermal amplification reactions, particularly in conjunction with the WarmStart LAMP Kit.
Catalog No. NC1640071 Supplier New England Biolabs, Inc. Supplier No. M1202S
Tte UvrD Helicase is a repair helicase capable of unwinding double-stranded DNA, without a requirement for a specific flap or overhang structure, from the thermophilic organism Thermoanaerobacter tengcongensis. It is active on a wide range of DNA substrates and, along with its thermostability (active to 70C), Tte UvrD Helicase has been demonstrated to be a useful additive for improving specificity of isothermal amplification reactions, particularly in conjunction with the WarmStart LAMP Kit.