Learn More
Bioworld TTE Buffer, Ready-Pack (20X), 1 PK(s)
Catalog No. 50198668
Encompass_Preferred
20x TTE (Tris-TAPS-EDTA) buffer pack can be dissolved into 2L of water to make 2L of 1x TTE solution.
Catalog No. 50-198-668 Supplier Bioworld Supplier No. 214200331
20x TTE (Tris-TAPS-EDTA) buffer pack can be dissolved into 2L of water to make 2L of 1x TTE solution.