Learn More
Bioworld TTETRIS-TAPS-EDTA) BUFFER
Catalog No. 50198665
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. TTE (TRIS-TAPS-EDTA) BUFFER POWDER BLEND: - Proteomics Grade - An electrophoresis running buffer for DNA Sequencing that yields longer reads
Catalog No. 50-198-665 Supplier Bioworld Supplier No. 214200311
This item has a minimum order quantity of 2 per supplier requirements. TTE (TRIS-TAPS-EDTA) BUFFER POWDER BLEND: - Proteomics Grade - An electrophoresis running buffer for DNA Sequencing that yields longer reads