Learn More
Biocare Medical, LLC TTF-1 + CK5 6ML
SDP
Supplier:  Biocare Medical, LLC PM425DSAA
Encompass_Preferred
TTF-1 + CK5 can aid the discrimination between lung adenocarcinoma (TTF-1) vs. lung SqCC (CK5). In most lung cancers tested, only a single antibody will stain. Co-expressions may be an indication of adenosquamous cell carcinomas.
Catalog No. 50-831-72